Skip to Content
Merck
All Photos(1)

Key Documents

890860P

Avanti

14:0 TAP

Avanti Research - A Croda Brand

Synonym(s):

1,2-dimyristoyl-3-trimethylammonium-propane (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$591.00

$591.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$591.00

About This Item

Empirical Formula (Hill Notation):
C34H68NO4Cl
CAS Number:
Molecular Weight:
590.36
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$591.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 200 mg (890860P-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[Cl-]

InChI

1S/C34H68NO4.ClH/c1-6-8-10-12-14-16-18-20-22-24-26-28-33(36)38-31-32(30-35(3,4)5)39-34(37)29-27-25-23-21-19-17-15-13-11-9-7-2;/h32H,6-31H2,1-5H3;1H/q+1;/p-1

InChI key

WOSYUIOSAOFINL-UHFFFAOYSA-M

General description

1,2-dimyristoyl-3-trimethylammonium-propane (14:0 TAP) is a saturated cationic lipid with a positively charged headgroup. It has an alkyl chain length with 14 carbon atoms.[1]

Application

1,2-dimyristoyl-3-trimethylammonium-propane (14:0 TAP) is suitable for:
  • the preparation of cationic liposomes[2]
  • reconstitution of bacteriorhodopsin (BR) and proteorhodopsin (PR) into liposomes[3]
  • composing lipid–styrene maleic acid (SMA) complexes[1]
  • liposome preparation by double emulsion method[4]

Biochem/physiol Actions

1,2-dimyristoyl-3-trimethylammonium-propane (14:0 TAP) is useful for bicelle formation. Bicelles can be integrated into standard crystallization protocols, and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation.

Packaging

5 mL Amber Glass Screw Cap Vial (890860P-200mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masafumi Tanaka et al.
Biochimica et biophysica acta. Biomembranes, 1862(5), 183209-183209 (2020-02-01)
Styrene maleic acid copolymers (SMA) form discoidal lipid nanoparticles (lipid nanodisks) that mimic plasma high-density lipoproteins. We have previously prepared and characterized lipid nanodisks composed of SMA and the neutral phospholipid 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC). In the present study, we tested whether
Lipid-induced dynamics of photoreceptors monitored by time-resolved step-scan FTIR spectroscopy
Jawurek M, et al.
Chemical Physics, 512, 53-61 (2018)
Reverse osmosis membranes based on a supported lipid bilayer with gramicidin A water channels
Saeki D, et al.
Desalination, 375, 48-53 (2015)
Nishu Kanwa et al.
Physical chemistry chemical physics : PCCP, 22(6), 3234-3244 (2020-01-30)
In the present contribution, we investigate the interactions of lipid bilayer membranes of different charges and different phase states with aliphatic amino acids of varying charge (aspartic acid, glutamic acid, arginine and lysine) and hydrophobicity (serine, leucine and valine) by
A E Regelin et al.
Biochimica et biophysica acta, 1464(1), 151-164 (2000-03-08)
In order to investigate the relationship between lipid structure and liposome-mediated gene transfer, we have studied biophysical parameters and transfection properties of monocationic DOTAP analogs, systematically modified in their non-polar hydrocarbon chains. Stability, size and (by means of anisotropy profiles)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service