Skip to Content
Merck
All Photos(4)

Key Documents

U3750

Sigma-Aldrich

Uridine

≥99%

Synonym(s):

1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
Beilstein:
754902
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

mp

163-167 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1

InChI key

DRTQHJPVMGBUCF-XVFCMESISA-N

Gene Information

mouse ... Uck1(22245)

Looking for similar products? Visit Product Comparison Guide

General description

Uridine is a uracil nucleoside, specific for RNA sequence. Uridine is present in blood, seminal fluid and cerebrospinal fluid. Uridine is synthesized in living organisms by de novo and salvage pathways.

Application

Uridine has been used:
  • as a component of hippocampal neuron medium for culture of primary hippocampal neuron cells
  • in uridine rescue experiments in mouse embryonic fibroblasts and human breast cancer cells
  • in chase sequencing experiments

Biochem/physiol Actions

Uridine monophosphate is essential for protein glycosylation, polysaccharide biosynthesis and lipid metabolism. Oral administration of uridine is suggested for anisopoikilocytosis and epileptic encephalopathy disorders. Uridine has numerous biological functions like treating dry eye syndrome, regulating nervous system and favors reproduction. High levels of uridine are implicated in insulin resistance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTEN regulates glutamine flux to pyrimidine synthesis and sensitivity to dihydroorotate dehydrogenase inhibition
Mathur D, et al.
Cancer Discovery, 7(4), 380390-380390 (2017)
Detection of phosphorylated Akt and MAPK in cell culture assays
Molgaard S, et al.
MethodsX, 3(2), 386398-386398 (2016)
Transient N-6-methyladenosine transcriptome sequencing reveals a regulatory role of m6A in splicing efficiency
Louloupi A, et al.
Cell Reports, 23(12), 34293437-34293437 (2018)
Biochemistry of uridine in plasma
Yamamoto T, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412(19-20), 17121724-17121724 (2011)
CAD mutations and uridine-responsive epileptic encephalopathy
Koch J, et al.
Brain, 140(2), 279286-279286 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service