Skip to Content
Merck
All Photos(2)

Key Documents

850337P

Avanti

MAPCHO-16

n-hexadecylphosphocholine, powder

Synonym(s):

HPC; Fos-Choline-16; FOS16; FC-16; Miltefosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$436.00
5 G
$1,690.00

$436.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$436.00
5 G
$1,690.00

About This Item

Empirical Formula (Hill Notation):
C21H46NO4P
CAS Number:
Molecular Weight:
407.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$436.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850337P-1g)
pkg of 1 × 5 g (850337P-5g)

manufacturer/tradename

Avanti Research - A Croda Brand 850337P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OCC[N+](C)(C)C)OCCCCCCCCCCCCCCCC

InChI

1S/C21H46NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-25-27(23,24)26-21-19-22(2,3)4/h5-21H2,1-4H3

InChI key

PQLXHQMOHUQAKB-UHFFFAOYSA-N

General description

Hexadecylphosphocholine is a structural analog of palmitoyl lysophosphatidylcholine that is more stable to hydrolytic degradation. The lipid is often used as a detergent for membrane protein solubilization and purification. It has been shown to have antineoplastic and antiprotozoal activity.

Application

MAPCHO-16 has been used to test against Leishmania intracellular amastigotes.[1] It has also been use to evaluate its efficacy in L. infantum chagasi infections.[2]

Biochem/physiol Actions

MAPCHO-16/ n-hexadecylphosphocholine/Miltefosine can be used as a stationary phase to determine total acidity of divalent cations by ion chromatography.[3] It is used in the oral treatment of human leishmaniasis [4]

Packaging

20 mL Clear Glass Screw Cap Vial (850337P-1g)
60 mL Amber Wide Mouth Glass Bottle with Screw Cap (850337P-5g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
MAPCHO is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 16-mercaptohexadecylphosphocholine, a miltefosine analog with leishmanicidal activity
Hornillos V, et al.
Bioorganic & medicinal chemistry letters, 16(19), 5190-5193 (2006)
Combination therapy with nitazoxanide and amphotericin B, Glucantime, miltefosine and sitamaquine against Leishmania (Leishmania) infantum intracellular amastigotes
Mesquita JT, et al.
Acta Tropica, 130, 112-116 (2014)
Determination of total acidity and of divalent cations by ion chromatography with n-hexadecylphosphocholine as the stationary phase
Hu W, et al.
Journal of Chromatography A, 956(1-2), 139-145 (2002)
Juliana Q Reimão et al.
PLoS neglected tropical diseases, 9(2), e0003556-e0003556 (2015-02-14)
The only oral drug available for the treatment of leishmaniasis is miltefosine, described and approved for visceral leishmaniasis in India. Miltefosine is under evaluation for the treatment of cutaneous leishmaniasis in the Americas although its efficacy for the treatment of
Bruno M Castro et al.
The journal of physical chemistry. B, 117(26), 7929-7940 (2013-06-07)
Edelfosine (1-O-octadecyl-2-O-methyl-sn-glycero-phosphocholine) and miltefosine (hexadecylphosphocholine) are synthetic alkylphospholipids (ALPs) that are reported to selectively accumulate in tumor cell membranes, inducing Fas clustering and activation on lipid rafts, triggering apoptosis. However, the exact mechanism by which these lipids elicit these events

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service