Skip to Content
Merck
All Photos(2)

Key Documents

850336P

Avanti

MAPCHO-12

Avanti Research - A Croda Brand

Synonym(s):

DPC; Fos-Choline-12; FOS12; FC-12

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$430.00
5 G
$1,650.00
25 G
$6,370.00

$430.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$430.00
5 G
$1,650.00
25 G
$6,370.00

About This Item

Empirical Formula (Hill Notation):
C17H38NO4P
CAS Number:
Molecular Weight:
351.46
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$430.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

n-dodecylphosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850336P-1g)
pkg of 1 × 25 g (850336P-25g)
pkg of 1 × 5 g (850336P-5g)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OCC[N+](C)(C)C)OCCCCCCCCCCCC

InChI

1S/C17H38NO4P/c1-5-6-7-8-9-10-11-12-13-14-16-21-23(19,20)22-17-15-18(2,3)4/h5-17H2,1-4H3

InChI key

QBHFVMDLPTZDOI-UHFFFAOYSA-N

General description

Dodecylphosphocholine is a structural analog of lauroyl lysophosphatidylcholine that is more stable to hydrolytic degradation. The lipid is often used as a detergent for membrane protein solubilization and purification.Dodecylphosphocholine has been shown to improve paracellular permeability of hydrophilic compounds by modulating the tight junctions. This may have implications on its use for oral or nasal drug delivery.
MAPCHO-12 or n-dodecylphosphocholine is a detergent with short chain.[1]

Application

MAPCHO-12 is suitable for use as detergent for B. subtilis cytochrome b558 protein for nuclear magnetic resonance experiments.[2] It is also suitable for use in the purification of cytoplasmic tail of gp41 (gp41CT) protein and micelle preparation.[3]

Biochem/physiol Actions

MAPCHO-12 or n-dodecylphosphocholine in combination with 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) is used for membrane-mimetic bicelle system for orientation based studies in membrane proteins.[1] It provides a zwitterionic surface in the membrane micelle structure.[4]

Packaging

125 mL Amber Wide Mouth Glass Bottle with Screw Cap (850336P-5g)
20 mL Clear Glass Screw Cap Vial (850336P-1g)
250 mL Amber Wide Mouth Glass Bottle with Screw Cap (850336P-25g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
MAPCHO is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetically oriented dodecylphosphocholine bicelles for solid-state NMR structure analysis
Nolandt OV, et al.
Biochimica et Biophysica Acta - Biomembranes, 1818(5), 1142-1147 (2012)
Nina Liu et al.
Heliyon, 5(12), e03018-e03018 (2020-01-01)
Fusion of host and viral membranes is a crucial step during infection by enveloped viruses. In the structurally-defined "class I″ viral glycoproteins, the formation of a highly stable α-helical bundle by the ectodomain of the fusion subunit (e.g., GP2 for
Thermodynamics, size, and dynamics of zwitterionic dodecylphosphocholine and anionic sodium dodecyl sulfate mixed micelles
Sikorska E, et al.
Journal of Thermal Analysis and Calorimetry, 123(1), 511-523 (2016)
Solution structure and membrane interaction of the cytoplasmic tail of HIV-1 gp41 protein
Murphy RE, et al.
Structure, 25(11), 1708-1718 (2017)
Sheeja V Vasudevan et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(15), 6847-6851 (2010-03-31)
Repair of damage to the central nervous system (CNS) is inhibited by the presence of myelin proteins that prevent axonal regrowth. Consequently, growth inhibitors and their common receptor have been identified as targets in the treatment of injury to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service