Skip to Content
Merck
All Photos(1)

Key Documents

800811O

Avanti

18:1 DG

Avanti Research - A Croda Brand

Synonym(s):

1-2-di-(9Z-octadecenoyl)-sn-glycerol; DG(18:1(9Z)/18:1(9Z)/0:0); DOG

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$161.00
25 MG
$284.00
200 MG
$780.00
500 MG
$1,530.00

$161.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$161.00
25 MG
$284.00
200 MG
$780.00
500 MG
$1,530.00

About This Item

Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$161.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

description

1-2-dioleoyl-sn-glycerol, neat oil

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 10 mg (with screw cap (800811O-10mg))
pkg of 2 × 100 mg (with screw cap (800811O-200mg))
pkg of 5 × 100 mg (with screw cap 800811O-500mg)
pkg of 1 × 25 mg (with screw cap (800811O-25mg))

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCC\C=C\CCCCCCCC)CO)C(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37,40H,3-16,21-36H2,1-2H3/b19-17+,20-18-/t37-/m0/s1

InChI key

AFSHUZFNMVJNKX-CSWDEDFYSA-N

General description

Diacylglycerol/diglyceride (DAG) is a glycerolipid, that is located at the plasma membrane.[1] It is made up of two fatty acid chains that are covalently attached to a single glycerol molecule with the help of an ester linkage.[2]

Application

18:1 DG has been used as a diacyl-glycerol source in diacylglycerol O-acyltransferase 1 (DGAT1) assay.[3] It has also been used as substrate to assess compounds as potential inhibitors of DGAT-1 in DGAT-1 enzyme assay.[4] It is suitable to use in lipid-protein overlay screen assay with the recombinant protein His-AtROP6.

Packaging

5 mL PTFE Vial with Screw Cap (800811O-10mg)
5 mL PTFE Vial with Screw Cap (800811O-200mg)
5 mL PTFE Vial with Screw Cap (800811O-25mg)
5 mL PTFE Vial with Screw Cap (800811O-500mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Probing the global cellular responses to lipotoxicity caused by saturated fatty acids
Piccolis M, et al.
Molecular Cell, 9, 347-347 (2019)
Identification of Inhibitors of triacylglyceride accumulation in muscle cells: comparing HTS results from 1536-well plate-based and high-content platforms
Sugarman E, et al.
Journal of Biomolecular Screening, 19(1), 77- 87 (2014)
Conical Lipids in Flat Bilayers Induce Packing Defects Similar to that Induced by Positive Curvature
Vamparys, L
Biophysical Journal, 104, 585-593 (2013)
Liposome-based carrier systems and devices used for pulmonary drug delivery
Biomaterials and Medical Tribology, 19(1), 395- 443 (2013)
Activation of ROP6 GTPase by phosphatidylglycerol in Arabidopsis
Han X, et al.
Frontiers in Plant Science, 9, 347-347 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service