Skip to Content
Merck
All Photos(2)

Key Documents

850104P

Avanti

18:0 Lyso PI

1-stearoyl-2-hydroxy-sn-glycero-3-phosphoinositol (ammonium salt), powder

Synonym(s):

1-octadecanoyl-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt); PI(18:0/0:0); 110719

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$284.00
500 μG
$1,130.00

$284.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$284.00
500 μG
$1,130.00

About This Item

Empirical Formula (Hill Notation):
C27H56NO12P
CAS Number:
Molecular Weight:
617.71
UNSPSC Code:
51191904
NACRES:
NA.25

$284.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

>99% (LPI; may contain up to 10% of the 2-LPI isomer, TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850104P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850104P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850104P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C27H53O12P.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(29)37-18-20(28)19-38-40(35,36)39-27-25(33)23(31)22(30)24(32)26(27)34;/h20,22-28,30-34H,2-19H2,1H3,(H,35,36);1H3/t20-,22-,23-,24+,25-,26-,27-;/m1./s1

InChI key

QVEDYTZLCBLSCH-FJYNCLFYSA-N

General description

Lysophosphatidylinositol (LPI), an acidic lysophosholipid,[1] carries inositol in its head group.[2]

Application

18:0 Lyso PI (1-stearoyl-2-hydroxy-sn-glycero-3-phosphoinositol) has been used as lysophosphatidylinositol (LPI) substrate in lysophosphatidylinositol acyltransferase biochemical assay to determine lyso-phosphatidylinositol acyltransferase (LPIAT) activity of liver microsomes.[3] It may be used as a synthetic LPI to compare its effects with liver LPI in transferrin (Tf) endocytosis.[4] It may also be used as LPI standard to quantify serum LPI levels in gonadectomized rats.[5]

Biochem/physiol Actions

Increased circulating levels of L-alpha-lysophosphatidylinositol (LPI) are associated with cancer and LPI is a potent ligand for the G-protein-coupled receptor GPR55.
Lysophosphatidylinositol (LPI) displays mitogenic activity and can serve as a bioactive lysophospholipid mediator.[2] It may be considered as a lipid mediator.[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850104P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850104P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Monica Imbernon et al.
Molecular and cellular endocrinology, 383(1-2), 159-169 (2014-01-01)
The G protein-coupled receptor GPR55 has been proposed as a new cannabinoid receptor associated with obesity in humans. We have investigated the regulation of GPR55 in rat white adipose tissue (WAT) in different physiological and pathophysiological settings involved in energy
Robert N Helsley et al.
eLife, 8 (2019-10-18)
Recent studies have identified a genetic variant rs641738 near two genes encoding membrane bound O-acyltransferase domain-containing 7 (MBOAT7) and transmembrane channel-like 4 (TMC4) that associate with increased risk of non-alcoholic fatty liver disease (NAFLD), non-alcoholic steatohepatitis (NASH), alcohol-related cirrhosis, and
Atsushi Yamashita et al.
Prostaglandins & other lipid mediators, 107, 103-116 (2013-05-30)
Lysophosphatidylinositol (LPI) is a subspecies of lysophospholipid and is assumed to be not only a degradation product of phosphatidylinositol (PI), but also a bioactive lysophospholipid mediator. However, not much attention has been directed toward LPI compared to lysophosphatidic acid (LPA)
Mariachiara Bianco et al.
Food chemistry, 324, 126878-126878 (2020-04-29)
Ceramides (Cer) and cerebrosides are important sphingolipids (SL) involved in many biological processes. Herein, the SL content of yellow lupin seeds (Lupinus luteus) was determined by liquid-liquid extraction, mild alkaline hydrolysis (1 h at 37 °C) and reversed-phase liquid chromatography with negative
Saori Oka et al.
Biochemical and biophysical research communications, 362(4), 928-934 (2007-09-04)
GPR55 is an orphan G protein-coupled receptor. In this study, we explored a possible endogenous ligand for GPR55 using HEK293 cells which expressed GPR55. We found that lysophosphatidylinositol induced rapid phosphorylation of the extracellular signal-regulated kinase in transiently or stably

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service