Skip to Content
Merck
All Photos(2)

Key Documents

858125C

Avanti

18:1 Lyso PG

1-oleoyl-2-hydroxy-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-oleoy-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(18:1(9Z)/0:0); 110714

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$357.00

$357.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$357.00

About This Item

Empirical Formula (Hill Notation):
C24H46O9PNa
CAS Number:
Molecular Weight:
532.58
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$357.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPG; may contain up to 10% of the 2-LPG isomer, TLC)

form

liquid

packaging

pkg of 5 × 4 mL (858125C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 858125C

concentration

25 mg/mL (858125C-500mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1 Lyso phosphoglycerol is a lyso-phospholipid detergent.[1] Lyso-phospholipids are produced in γ-proteobacteria by lipoprotein acyltransferase activity. These lipids are found in minimal amount in normal bacterial membranes.[2]

Application

18:1 Lyso PG has been used as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS) method conditions.[3]

Biochem/physiol Actions

Lyso phosphoglycerol (Lyso PG) and acyl-CoA acts as a substrate for lysophosphatidylglycerol acyltransferase 1 (LPGAT1) to synthesize phosphoglycerol.[4] Lyso-phospholipids are nonbilayer-forming lipids. Accumulation of lysophospholipids in bacterial membranes disrupt the membrane integrity due to its detergent- like physical property.[2]

Packaging

5 mL Clear Glass Sealed Ampule (858125C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression, purification and reconstitution of the C-terminal transmembrane domain of scavenger receptor BI into detergent micelles for NMR analysis
Chadwick A, et al.
Protein Expression and Purification, 107, 35-42 (2015)
Substrate selectivity of lysophospholipid transporter LplT involved in membrane phospholipid remodeling in Escherichia coli
Lin Y, et al.
The Journal of Biological Chemistry, 291(5), 2136-2149 (2016)
Comprehensive untargeted lipidomic analysis using core-shell C30 particle column and high field orbitrap mass spectrometer
Narvaez-Rivas M and Zhang Q
Journal of Chromatography A, 1440, 123-134 (2016)
Hideo Shindou et al.
The Journal of biological chemistry, 284(1), 1-5 (2008-08-23)
Cell membranes contain several classes of glycerophospholipids, which have numerous structural and functional roles in the cells. Polyunsaturated fatty acids, including arachidonic acid and eicosapentaenoic acid, are located at the sn-2 (but not sn-1)-position of glycerophospholipids in an asymmetrical manner.
Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service