Skip to Content
Merck
All Photos(2)

Key Documents

840434P

Avanti

10:0 PG

1,2-didecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

PG(10:0/10:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$253.00

$253.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$253.00

About This Item

Empirical Formula (Hill Notation):
C26H50O10PNa
CAS Number:
Molecular Weight:
576.63
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$253.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840434P-25mg)
pkg of 1 × 500 mg (840434P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840434P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCCCC)=O)COC(CCCCCCCCC)=O.[Na+]

InChI

1S/C26H51O10P.Na/c1-3-5-7-9-11-13-15-17-25(29)33-21-24(22-35-37(31,32)34-20-23(28)19-27)36-26(30)18-16-14-12-10-8-6-4-2;/h23-24,27-28H,3-22H2,1-2H3,(H,31,32);/q;+1/p-1/t23?,24-;/m1./s1

InChI key

WOGMIDKKARMDEU-RETUOTIWSA-M

General description

Phosphatidylglycerol (PG) is one of the important lipid, found in the membranes of bacteria. But in mammals, PG accounts for only 1–2 mol% of phospholipids. This lipid is abundantly present in the lung surfactant. thus pointing to an important role therein.[1]

Application

10:0 PG (1,2-didecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) may be used as a component in the liposomes, labeled with β-pyrene-C10-PG.[2] It may also be used in the preparation of small unilamellar vesicle (SUV) liposomes.[3]

Biochem/physiol Actions

Phosphatidylglycerol (PG) serves as an indicator of lung maturity in fetus.[4]

Packaging

30 mL Amber Wide Mouth Bottle with Screw Cap (840434P-500mg)
5 mL Clear Glass Sealed Ampule (840434P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 223-249 (2008)
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 223-249 (2008)
Alison H Dewald et al.
Biophysical journal, 100(9), 2131-2140 (2011-05-05)
The spontaneous folding of two Neisseria outer membrane proteins, opacity-associated (Opa)(60) and Opa(50) into lipid vesicles was investigated by systematically varying bulk and membrane properties. Centrifugal fractionation coupled with sodium dodecyl sulfate polyacrylamide gel electrophoresis mobility assays enabled the discrimination
Camilo F Muñoz et al.
Frontiers in plant science, 10, 1573-1573 (2019-12-19)
Microalgal lipids are promising feedstocks for food and biofuels. Since lipid production by microalgae is not yet economically feasible, genetic engineering is becoming a promising strategy to achieve higher lipid accumulation and productivities. Enzymes involved in the Kennedy pathway such
Brianna M Abbott et al.
Biochimica et biophysica acta. Biomembranes, 1860(5), 1187-1192 (2018-02-13)
Experiments investigating the adsorption and desorption of cytochrome c onto and from liposomes containing 50 mol% 1,2-diacylphosphatidylglycerol lipids [10:0, 12:0, 14:0, 16:0, 18:1(Δ9 cis)] with 1,2-dioleoyl-sn-glycero-3-phosphatidylcholine (DOPC) in pH 7.4 buffered solutions of low to moderate ionic strength are reported. Fluorescence experiments

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service