Skip to Content
Merck
All Photos(2)

Documents

840060P

Avanti

16:0-18:2 PS

1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-L-serine (sodium salt), powder

Synonym(s):

1-hexadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phospho-L-serine (sodium salt); PS(16:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H73NO10PNa
CAS Number:
Molecular Weight:
781.97
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840060P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840060P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H74NO10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(43)51-36(34-49-52(46,47)50-35-37(41)40(44)45)33-48-38(42)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,36-37H,3-10,12,14-16,19-35,41H2,1-2H3,(H,44,45)(H,46,47);/q;+1/p-1/b13-11-,18-17-;/t36-,37+;/m1./s1

InChI key

WPLLCKLCLMRAPV-WHVRKWPVSA-M

General description

Phosphatidylserine (PS) is an acidic phospholipid, abundantly found in the protein myelin. It is localized to the membranes and present in plasma. PS constitutes 2% to 20% of the total phospholipid mass.

Application

16:0-18:2 PS or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-L-serine (sodium salt) might be used in the preparation of giant unilamellar liposomes (GULs).

Biochem/physiol Actions

Phosphatidylserine (PS) supports nerve cell membranes and myelin. It affects the catabolism and anabolism of neurotransmitters acetylcholine (ACh), norepinephrine, serotonin and dopamine.

Packaging

5 mL Clear Glass Sealed Ampule (840060P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Y Dunina-Barkovskaya et al.
The Journal of membrane biology, 198(2), 77-87 (2004-05-13)
Low gadolinium concentrations induce rapid gigaseal formation and cell adhesion to glass and plastic (polystyrene) substrates in the slime mutant of Neurospora crassa. Cellular adhesion is independent of an integrin-mediated mechanism, because pretreatment with the oligopeptide ARG-GLY-ASP-SER (RGDS) did not
Michael J Glade et al.
Nutrition (Burbank, Los Angeles County, Calif.), 31(6), 781-786 (2015-05-03)
The aim of this study was to assess the roles and importance of phosphatidylserine (PS), an endogenous phospholipid and dietary nutrient, in human brain biochemistry, physiology, and function. A scientific literature search was conducted on MEDLINE for relevant articles regarding
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service