Skip to Content
Merck
All Photos(2)

Documents

850759C

Avanti

16:0-20:4 PE

1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-hexadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphoethanolamine; PAPE; PE(16:0/20:4(5Z,8Z,11Z,14Z)); 110638

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H74NO8P
CAS Number:
Molecular Weight:
740.00
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 2.5 mL (850759C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850759C

concentration

10 mg/mL (850759C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

Application

16:0-20:4 PE has been used to determine its effect on oxidative burst in neutrophil granulocytes. It has also been used to determine its detergent solubility with equimolar concentration of sphingomyelin/cholesterol (SM/CHOL).

Packaging

5 mL Clear Glass Sealed Ampule (850759C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The oxidation state of phospholipids controls the oxidative burst in neutrophil granulocytes
Bluml S, et al.
Journal of Immunology, 181(6), 4347-4353 (2008)
Acyl chain unsaturation in PEs modulates phase separation from lipid raft molecules
Shaikh SR, et al.
Biochemical and Biophysical Research Communications, 311(3), 793-796 (2003)
Bruna Neves et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-07)
Nitrated phospholipids have recently been detected in vitro and in vivo and associated with beneficial health effects. They were identified and quantified in biological samples by lipidomics methodologies using liquid chromatography-collision-induced dissociation (CID) tandem mass spectrometry (MS/MS) acquired with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service