Skip to Content
Merck
All Photos(2)

Key Documents

810118C

Avanti

Egg NBD PE

Avanti Research - A Croda Brand 810118C

Synonym(s):

L-α-Phosphatidylethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (Ammonium Salt) (Egg-Transphosphatidylated, Chicken)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$375.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$375.00

About This Item

Empirical Formula (Hill Notation):
C45H80N5O11P
CAS Number:
Molecular Weight:
898.12
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810118C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810118C

concentration

1 mg/mL (810118C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

The phosphatidylethanolamine (PE) is a zwitterionic phospholipid. Its headgroup is provided with a phosphate, that is negatively charged and a free amine, that is positively charged.[1] PE is one of the widely distributed glycerophospholipid in eukaryotic cells.[2]

Application

Egg NBD PE is suitable to use as an additive in dimethylformamide (DMF) to compare the effects of nystatin on the diffusion of NBD-Nys and NBD-PE.[3] It has also been used to monitor lipid extraction from liposomes for lipid transfer assay.[4]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) helps to activate factor (F)X and PT, thereby stimulating thrombosis. It serves as an anticoagulant.[5] PE is essential for the surface expression of γ-aminobutyric acid type A receptors (GABAARs) in cultured neurons.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (810118C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibodies and Diagnostic Tests in Antiphospholipid Syndrome
Systemic Lupus Erythematosus, 495- 501 (2016)
Proteostasis maintenance of Cys-loop receptors
Advances in Protein Chemistry and Structural Biology, 103(3), 1- 23 (2016)
Evidence for nystatin micelles in L-cell membranes from fluorescence photobleaching measurements of diffusion
O'Neill LJ, et al.
Biochemistry, 25(1), 177- 181 (1986)
Phosphatidylethanolamine metabolism in health and disease
International Review of Cell and Molecular Biology, 321(3), 29- 88 (2016)
Lipid-Protein Interactions in Membranes
Yeagle PL
The Membranes of Cells, 291- 334 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service