Skip to Content
Merck
All Photos(1)

Key Documents

880132P

Avanti

DSPE-PEG(5000) Amine

Avanti Research - A Croda Brand, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$446.00

$446.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$446.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$446.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (880132P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH3)113]N)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

General description

DSPE-PEG(5000) Amine is a functionalized polyethylene glycol (PEG) lipid, containing two stearic acid as a hydrophobic moiety.[1]

Application

DSPE-PEG(5000) Amine has been used in the preparation of polyethylene glycol (PEG)-lipid monolayers for neutron reflectivity study.[2]

Packaging

5 mL Clear Glass Sealed Ampule (880132P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced stability of PEG-block-poly (N-hexyl stearate l-aspartamide) micelles in the presence of serum proteins
Diezi TA, et al.
Molecular Pharmaceutics, 7(4), 1355-1360 (2010)
Folate-targeted gadolinium-lipid-based nanoparticles as a bimodal contrast agent for tumor fluorescent and magnetic resonance imaging
Nakamura T, et al.
Biological & Pharmaceutical Bulletin, 37(4), 521-527 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service