Skip to Content
Merck
All Photos(1)

Key Documents

870160P

Avanti

16:0 Ptd Thioethanol

Avanti Research - A Croda Brand 870160P, powder

Synonym(s):

1,2-Dipalmitoyl-sn-Glycero-3-Phosphothioethanol (Sodium Salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$230.00
100 MG
$619.00
500 MG
$1,370.00

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$230.00
100 MG
$619.00
500 MG
$1,370.00

About This Item

Empirical Formula (Hill Notation):
C37H72O8PNaS
CAS Number:
Molecular Weight:
730.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 g (870160P-1g)
pkg of 1 × 100 mg (870160P-100mg)
pkg of 1 × 25 mg (870160P-25mg)
pkg of 1 × 500 mg (870160P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870160P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCS)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H73O8PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(38)42-33-35(34-44-46(40,41)43-31-32-47)45-37(39)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35,47H,3-34H2,1-2H3,(H,40,41);/q;+1/p-1/t35-;/m1./s1

InChI key

AKETWRINAWSUQU-RUQJKXHKSA-M

General description

1,2-Dipalmitoyl-sn-Glycero-3-Phosphothioethanol (DPPTE) or 16:0 Ptd Thioethanol is a thiol modified phospholipid.[1]

Application

16:0 Ptd Thioethanol has been used in the preparation of unilamellar liposomes[2] and supported lipid bilayers (SLB).[2][1]

Biochem/physiol Actions

1,2-Dipalmitoyl-sn-Glycero-3-Phosphothioethanol (DPPTE) or 16:0 Ptd Thioethanol facilitates the liposome formation by building the lipid layers with the assistance of thiol-anchors at the lipid headgroups.[1]

Packaging

20 mL Clear Glass Screw Cap Vial (870160P-1g)
20 mL Clear Glass Screw Cap Vial (870160P-500mg)
5 mL Amber Glass Screw Cap Vial (870160P-100mg)
5 mL Amber Glass Screw Cap Vial (870160P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of thiolated phospholipids on formation of supported lipid bilayers on gold substrates investigated by surface-sensitive methods
Kilicc, A, et al.
Colloids and Surfaces. B, Biointerfaces, 160, 117-125 (2017)
Peptide-functionalized supported lipid bilayers to construct cell membrane mimicking interfaces
Kilic A and Kok FN
Colloids and Surfaces. B, Biointerfaces, 176, 18-26 (2019)
Lena A Lautscham et al.
Biomaterials, 35(10), 3198-3207 (2014-01-21)
Cell behavior such as cell adhesion, spreading, and contraction critically depends on the elastic properties of the extracellular matrix. It is not known, however, how cells respond to viscoelastic or plastic material properties that more closely resemble the mechanical environment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service