Z743032
Julabo® CORIO™ CD heating circulator
CD-BC6, bath capacity 4.5 L (6 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, Schuko plug
All Photos(1)
About This Item
Recommended Products
material
stainless steel bath
manufacturer/tradename
Julabo 9012506
temperature measuring range
20-150 °C
bath capacity
4.5 L (6 L)
external W × L × H
24 cm × 44 cm × 47 cm
opening W × L × H
13 cm × 15 cm × 20 cm
General description
CORIO CD heating circulators feature professional technology for high standards. These units facilitate internal temperature control in the bath tank or control of externally connected applications.
Your advantages
Your advantages
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy pump change-over between internal and external circulation
- USB connection
- High-quality bath tanks made of stainless steel with bath lid and drain tap
- Integrated cooling coil for counter-cooling
Legal Information
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.