Key Documents
SYNH200001
Synthware aluminum heat transfer block for mini vials
Select a Size
1 EA
$159.00
$159.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$159.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.51
$159.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
H200001
Kemtech H200001
General description
Synthware aluminum heat transfer block is used in place of a glass sand bath in microscale experiments. It has holes to hold 3.0 mL and 5.0 mL heavy-wall reaction vials, as well as four Craig crystallization tubes, with one holder on the top for a standard thermometer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)