Skip to Content
Merck
All Photos(1)

Key Documents

Z742926

Citrajet® detergent, low foam liquid acid cleaner

pkg of 3.8 L

Synonym(s):

biodegradable cleaner, low foaming acid detergent, metal oxide remover, phosphate-free cleaner, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing

Select a Size

3.8 L
$108.00
4 X 3.8 L
$362.00

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
3.8 L
$108.00
4 X 3.8 L
$362.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pkg of 3.8 L

manufacturer/tradename

Alconox, Inc.

Looking for similar products? Visit Product Comparison Guide

General description

Citrajet® is a low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high peformance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing. Dilute 2:100 pH 2.5

Features and Benefits

  • concentrated
  • ideal for use as acid wash or rinse
  • phosphate free, biodegradable and readily disposable
  • combines acid and detergent ingredients to remove metal oxides, salts and inorganic residues

Legal Information

Citrajet is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service