BR146508
BRAND® glass powder funnel short wide stem
top diam. 60 mm, bottom diam. 18 mm
Synonym(s):
Funnels, Power funnels
Select a Size
$16.70
Select a Size
About This Item
$16.70
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
BRAND 146508
bottom diam.
18 mm
top diam.
60 mm
Related Categories
General description
Features and Benefits
- BRAND® borosilicate glass powder funnel is best suited for transferring powders.
- These general lab use funnels include ground and fire-polished upper rim.
- Features a short, wide stem that facilitates optimal filtration.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)