Skip to Content
Merck
All Photos(1)

Documents

Z742927

Citrajet® detergent, low foam liquid acid cleaner

pkg of 19 L Jerrycan

Synonym(s):

biodegradable cleaner, low foaming acid detergent, metal oxide remover, phosphate-free cleaner, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

packaging

pkg of 19 L Jerrycan

manufacturer/tradename

Alconox, Inc.

Looking for similar products? Visit Product Comparison Guide

General description

Citrajet® is a low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high peformance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing. Dilute 2:100 pH 2.5

Features and Benefits

  • concentrated
  • ideal for use as acid wash or rinse
  • phosphate free, biodegradable and readily disposable
  • combines acid and detergent ingredients to remove metal oxides, salts and inorganic residues

Legal Information

Citrajet is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service