Key Documents
Z742419
Q-Tube gas purging set
12 SS, pack of 1 ea
Select a Size
1 EA
$2,380.00
$2,380.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$2,380.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.51
$2,380.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Q Labtech 100-QT00PG-012SS
parameter
180 psi max. pressure
General description
The parts with direct chemical contact are made of 316 stainless steel, PTFE, Simriz, or glass.
Q-Tube-Purging-12-SS / 100-QT00PG-012SS can be operated safely and accurately with high reproducibility with a wide range of gases, including HCl, NH3, H2, O2, N2, Ar, D2, CO, CO2, and etc. The set contains all items needed for immediate use.
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)