Key Documents
Z740135
QianCap pressure monitor-25
for tubes/bottles with internal threads #25
Select a Size
1 EA
$1,540.00
$1,540.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,540.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$1,540.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Q Labtech
parameter
0-300 psi pressure range
Related Categories
General description
QianCap pressure monitor-25 is designed for monitoring internal pressure on pressure tubes, flasks and bottles with #25 internal threads. The set contains all items needed for immediate use (pressure release system, 1 PTFE adapter-25H, 1 Simriz O-ring-25-S, 1 stainless steel pressure gauge-300 psi and 10 septa-12 mL).
Features and Benefits
  • Monitors Ace-Thred pressure tubes/bottles.
  • Provides optional higher pressure adapter (120 psi).
Legal Information
QianCap is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)