Key Documents
Z682128
Ace cold trap outer body only for condenser "C" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
Select a Size
1 EA
$2,150.00
$2,150.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$2,150.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$2,150.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
glass
manufacturer/tradename
Büchi equivalent, 40641
Related Categories
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
Z682071Z682292Z682136
manufacturer/tradename
Büchi equivalent, 40641
manufacturer/tradename
Büchi equivalent, 40643
manufacturer/tradename
Büchi equivalent, 11228
manufacturer/tradename
Büchi equivalent, 00672
General description
The cold trap outer body for the Ace condenser "C" assembly, is glass with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks. Features standard top and side flange that connects directly to the Büchi rotary evaporator.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)