Key Documents
Z682055
Ace rotary evaporator condenser complete "C" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
Select a Size
1 EA
$3,900.00
$3,900.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$3,900.00
About This Item
UNSPSC Code:
41121800
NACRES:
VB.02
$3,900.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
plastic-coated glass
manufacturer/tradename
Büchi equivalent, 40646
Related Categories
General description
The "C" condenser assembly consists of the main glass outer trap body with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks.
Features standard top and side flange that connects directly to the Büchi® rotary evaporator. The assembly also includes the glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)