Key Documents
Z645230
IKA® M 23 star-shaped cutting blade
M 23 star-shaped cutter, 1/cs
Select a Size
1 EA
$853.00
$853.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$853.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
$853.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
stainless steel
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 1443400
suitability
suitable for
Related Categories
General description
stainless steeel. star knife for use with the IKA M 20 Universal Mill. For shredding fibrous materials such as paper and plant material, but also for plastics and other materials with a low specific weight.
used to crush fibrous substances such as paper and vegetation, but also for plastics and material with a low specific weight.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)