Key Documents
Z645222
IKA® M 22 hard metal cutter
M 22 hard metal cutter, tungsten carbide, 1/cs
Select a Size
1 EA
$1,050.00
$1,050.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,050.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
$1,050.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
tungsten carbide
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 521800
suitability
suitable for (hard materials up to Mohs hardness 9)
General description
Note: Made of tungsten carbide for hard materials up to Mohs hardness 9.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. is this blade meant to work with the IKA M20 universal mill?
    1 answer
    1. This blade is compatible with the IKA M 20 Universal Mill.
      Helpful?