Key Documents
Z615595
Julabo® bath carrier trays for SW22/SW23
to fit 32 Erlenmeyer flasks 50 cc
Select a Size
1 EA
$638.00
$638.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$638.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.77
$638.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
Julabo 8970361
Julabo
General description
Erlenmeyer flasks can be positioned in the shaking water bath using the stainless steel tray. A maximum of 32 Erlenmeyer flasks with 50 ml capacity fit on a standard tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)