Skip to Content
Merck
All Photos(1)

Key Documents

Z156485

Sigma-Aldrich

O-ring set

O-ring set

Viton®

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$860.00

$860.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
1 SET
$860.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

$860.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


material

Viton®

manufacturer/tradename

Ace Glass

Looking for similar products? Visit Product Comparison Guide

General description

500 O-rings in a storage box. Thirty sizes ranging from: 1/8 I.D. × 1/4 in. O.D. up to 1 3/4 I.D. × 2 1/8 in. O.D. Use nitrile for economical sealing, Viton® for thermal or chemical resistance.

Legal Information

Viton is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Please specify the diameter of all set of O rings..

    1 answer
    1. This O-Ring set includes the items listed below. The O-Ring size is followed by the internal diameter (I.D.).

      35 each with a width of 1.78mm: -006/2.9mm, -007/3.7mm, -008/4.5mm, -009/5.3mm, -010/6.1mm, -011/7.7mm, -012/9.2mm
      20 each with a width of 2.6mm: -110/9.2mm, -111/10.8mm, -112/12.4mm, -113/13.9mm
      16 each with a width of 2.6mm: -114/15.5mm, -115/17.1mm, -116/18.7mm
      10 each with a width of 3.5mm: -210/18.6mm, -211/20.2mm, -212/21.8mm, -213/23.4mm, -214/25.0mm, -215/26.6mm, -216/28.2mm
      7 each with a width of 3.5mm: -217/29.7mm, -218/31.3mm, -219/32.9mm, -220/34.5mm, -221/36.1mm, -222/37.7mm
      5 each with a width of 5.0mm: -325/37.5mm, -326/40.6mm, -327/43.8mm

      Helpful?

  2. What are the dimensions of the O-Rings?

    1 answer
    1. The set contains 500 O-rings with the following sizes:
      • 35 each: -006, -007, -008, -009, -010, -011, -012
      • 20 each: -110, -111, -112, -113
      • 16 each: -114, -115, -116
      • 10 each: -210, -211, -212, -213, -214, -215, -216
      • 7 each: -217, -2

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service