Skip to Content
Merck
All Photos(2)

Key Documents

HS23263A

Vertical storage rack

to hold, 8 x 80 well racks, clear acrylic rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$235.00

$235.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
1 EACH
$235.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.15

$235.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


material

clear acrylic rack

feature

to hold 8 x 80 well racks

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific HS23263A

L × W × H

157 mm × 236 mm × 234 mm

General description

The Vertical Storage Rack for 80-wells is strong, clear, and of acrylic construction. These racks help you keep large numbers of microtubes organized on your benchtop.

Features and Benefits

  • Holds up to eight 80-well microtube racks filled with either 1.5 or 2.0 mL microtubes
  • Plenty of room to slide the racks in and out

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service