Key Documents
Z569658
KDS syringe multi-rack for Legato series pumps
small
Synonym(s):
dosing pump syringe rack, infusion pump syringe rack
Select a Size
1 EA
$1,270.00
$1,270.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,270.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
$1,270.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
KD Scientific 78-8300
General description
Accommodates up to 6 x 30 to 60 mL syringes or up to ten 0.5 μL to 20 mL. Works with Legato 200, 210 and 210P.
Legal Information
Legato is a trademark of KD Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)