Key Documents
Z568759
Simriz® high-performance O-ring for QianCap
25-S
Select a Size
1 EA
$113.00
$113.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$113.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$113.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
Q Labtech
General description
Have excellent compatibility with most of the chemicals, including inorganic and organic acids and bases, alcohols, aliphatic and chlorinated hydrocarbons, aromatics, ketones, esters, ethers and aldehydes and fuel oils.
Legal Information
QianCap is a trademark of Q Labtech LLC
Simriz is a registered trademark of Carl Freudenburg KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)