Key Documents
Z147958
Sigma-Aldrich
Aldrich® reverse-flow bubbler
capacity 25 mL , per chamber
Synonym(s):
bubblers, scientific bubblers
Select a Size
1 EA
$328.00
$328.00
Estimated to ship onMay 29, 2025
Select a Size
Change View
1 EA
$328.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$328.00
Estimated to ship onMay 29, 2025
capacity
25 mL , per chamber
General description
Monitors gas flow into, or evolution from a system.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)