Key Documents
Z549622
Sigma-Aldrich
Stopcock Adapter
PTFE side-port stopcock, joint: ST/NS 24/40
Select a Size
1 EA
$143.00
$143.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$143.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$143.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PTFE side-port stopcock
Joint
joint: ST/NS24/40
Related Categories
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)