Key Documents
Z549509
Sigma-Aldrich
Stopcock Adapter
straight glass stopcock, joint: ST/NS 29/32
Select a Size
1 EA
$129.00
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$129.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
straight glass stopcock
Joint
joint: ST/NS29/32
Related Categories
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)