Key Documents
Z176044
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter without return hole
female joint: ST/NS 24/40, male joint: ST/NS 24/40, capacity 100 mL
Select a Size
1 EA
$215.00
$215.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$215.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$215.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Joint
female joint: ST/NS24/40
male joint: ST/NS24/40
capacity
100 mL
Related Categories
General description
Aldrich® antisplash splash-guard adapter without return hole are traps that greatly reduce carry-over from foaming or bumping. Whatever comes up into this trap will stay and not go back to the evaporator flask.
Application
Aldrich® antisplash splash-guard adapter without return hole is ideal for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping".
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)