Skip to Content
Merck
All Photos(1)

Key Documents

Z317594

Duran® vacuum desiccator plate

porcelain plate, I.D. 300 mm

Synonym(s):

297256909

Sign Into View Organizational & Contract Pricing

Select a Size

Z317594-1EA
$371.00

$371.00


Estimated to ship onMarch 30, 2025



Select a Size

Change View
Z317594-1EA
$371.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
material:
porcelain plate
I.D.:
300 mm

$371.00


Estimated to ship onMarch 30, 2025


material

porcelain plate

manufacturer/tradename

DURAN®
Schott

availability

not available in USA and Canada

I.D.

300 mm

Looking for similar products? Visit Product Comparison Guide

General description

Perforated plate for vacuum desiccators.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service