Key Documents
Z291668
Supelco
Screw cap (open-top), polypropylene
black polypropylene, open-top, 8-425 thread, pkg of 100 ea
Select a Size
100 EA
$26.90
$26.90
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Request a Bulk Order
Select a Size
Change View
100 EA
$26.90
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
$26.90
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Request a Bulk Order
material
black polypropylene
Agency
suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
reg. compliance
suitable for FDA C-010.02
feature
autoclavable
closure type black polypropylene hole cap
packaging
pkg of 100 ea
parameter
135 °C temp. limit
fitting
8-425 thread
application(s)
PFAS testing
General description
These hole caps do not come with septa.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)