Key Documents
Z203335
Sigma-Aldrich
Aldrich® vacuum-transfer adapter
joint: ST/NS 29/32
Select a Size
1 EA
$415.00
$415.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$415.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$415.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS29/32
stopcock size
4 mm
Related Categories
General description
PTFE stopcocks. For transfer of hazardous and sensitive reagents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)