Key Documents
Z202789
Sigma-Aldrich
Aldrich® adjustable rapid-purge valve
About This Item
UNSPSC Code:
40141600
NACRES:
NB.79
Pricing and availability is not currently available.
Related Categories
General description
Designed for mounting anywhere in a vacuum system. Use for purging evacuated apparatus with inert gas. Two-piece PTFE plug with Viton® O-ring and SS spring is adjustable for pressures of 5 to 35 psi.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)