Key Documents
SYNS620012
Synthware 180 degree high vacuum valve with protected o-rings
size 0-12 mm
Synonym(s):
Synthware S620012
Select a Size
1 EA
$144.00
$144.00
Estimated to ship onMay 23, 2025
Select a Size
Change View
1 EA
$144.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$144.00
Estimated to ship onMay 23, 2025
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S620012
Related Categories
General description
With two 9 mm O.D. sidearms sealed to the barrel at a 180°angle to one another. Viton® O-ring on PTFE shaft are protected with precision PTFE wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal. This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)