Skip to Content
Merck
All Photos(1)

Documents

341276

Sigma-Aldrich

Anti-Factor H Goat pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

availability

not available in Canada

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CFH(3075)

General description

Anti-Factor H, goat polyclonal, recognizes Factor H in human. It is validated for Radial Immunodiffusion and Ouchterlony.
Goat polyclonal antibody supplied as undiluted serum. Recognizes Factor H.
Recognizes Factor H.

Immunogen

Human
purified, human factor H

Application

Radial Immunodiffusion (see comments)

Ouchterlony (≥ undiluted)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

This antibody has also been reported to work for radial immunodiffusion. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Ihnatko et al.
Frontiers in medicine, 8, 789972-789972 (2022-01-22)
COVID-19 vaccine-induced thrombotic thrombocytopenia (VITT) is a rare complication of adenoviral vector (ChAdOx1 nCoV-19) vaccine administration. It is presented as thrombocytopenia and thrombotic manifestations in various sites, especially in cerebral veins. Pulmonary emboli have been reported rarely. We present a
Anuj Pathak et al.
Nature communications, 9(1), 3398-3398 (2018-08-25)
Streptococcus pneumoniae evades C3-mediated opsonization and effector functions by expressing an immuno-protective polysaccharide capsule and Factor H (FH)-binding proteins. Here we use super-resolution microscopy, mutants and functional analysis to show how these two defense mechanisms are functionally and spatially coordinated
Jingyi Li et al.
Frontiers in immunology, 15, 1368322-1368322 (2024-04-01)
Activation of complement through the alternative pathway (AP) has a key role in the pathogenesis of IgA nephropathy (IgAN). We previously showed, by intraperitoneal injection of Lactobacillus casei cell wall extract (LCWE), C57BL/6 mice develop mild kidney damage in association
Federica Bianchi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(11), 12099-12111 (2019-08-24)
The 4 component meningococcus B vaccine (4CMenB) vaccine is the first vaccine containing recombinant proteins licensed for the prevention of invasive meningococcal disease caused by meningococcal serogroup B strains. 4CMenB contains 3 main recombinant proteins, including the Neisseria meningitidis factor
Diana Pauly et al.
Cell reports, 29(9), 2835-2848 (2019-11-28)
Complement dysregulation is a feature of many retinal diseases, yet mechanistic understanding at the cellular level is limited. Given this knowledge gap about which retinal cells express complement, we performed single-cell RNA sequencing on ∼92,000 mouse retinal cells and validated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service