Skip to Content
Merck
All Photos(1)

Key Documents

Z174440

Sigma-Aldrich

Aldrich® short-path distillation apparatus

Standard head

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,450.00

$1,450.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$1,450.00

About This Item

UNSPSC Code:
41104812
NACRES:
NB.43

$1,450.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Threaded 24/40 joints. 100 mL two-neck flask and 50 mL recovery flask.

Replacement parts:
Z174459 - Standard head
Z247839 - Jacketed head
Z174491 - Receiver flask, 50 mL
Z174483 - Two-neck flask, 100 mL
Z405531 - Two-neck flask, 250 mL
Z174475 - Gas-inlet adapter
Z174467 - Gas-inlet tube

Please note: Purchase thermometer separately (Z252980)

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service