Key Documents
SYND251014
Synthware Vigreux microscale distillation head
joint: ST/NS 14/20, immersion depth 25 mm
Synonym(s):
Synthware D251014, short path, distilling head
Select a Size
1 EA
$233.00
$233.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$233.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
$233.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D251014
immersion depth
25 mm
Related Categories
General description
With Vigreux Indentations, used with 25 mm immersion thermometer, and D27 distillation receiver.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)