Key Documents
Z173398
Sigma-Aldrich
Aldrich® Schlenk-type adapter
Joint cap with stopcock, female joint: threaded 14/20
Select a Size
1 EA
$134.00
$134.00
Estimated to ship onMay 29, 2025
Select a Size
Change View
1 EA
$134.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$134.00
Estimated to ship onMay 29, 2025
Joint
female joint: threaded14/20
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)