Key Documents
SYNA722440
Synthware metering flow control adapter with PTFE stopcock
joint: ST/NS 14/20
Synonym(s):
Synthware A722440, flow control adapter
Select a Size
1 EA
$245.00
$245.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$245.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$245.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A722440
Related Categories
General description
Provides on/off control with the main valve. Allows delivery of preset volumes of gases or liquids to reactions using the needle valve.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)