Key Documents
Z169463
Supelco
Spectroline® E-Series lamp bulb
shortwave, output6 W
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
Pricing and availability is not currently available.
technique(s)
thin layer chromatography (TLC): suitable
output
6 W
Application
Spectroline E-Series lamp bulb was used for illumination of trans-bis(trimethylammoniumhexyloxy)azobenzene dibromide solution to produce solution of its cis isomer†.[1]
Legal Information
Spectroline is a registered trademark of Spectronics, Corp.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Photocrosslinkable modified vegetable oil based resin for wood surface coating application
Mahendran, Arunjunai Raj, et al.
Progress in Organic Coatings, 74, 697-704 (2012)
F Pierce Hubbard et al.
Langmuir : the ACS journal of surfaces and colloids, 23(9), 4819-4829 (2007-03-27)
We report light and small-angle neutron scattering measurements that characterize microstructures formed in aqueous surfactant solutions (up to 1.0 wt % surfactant) containing mixtures of sodium dodecyl sulfate (SDS) and the light-sensitive bolaform surfactant, bis(trimethylammoniumhexyloxy)azobenzene dibromide (BTHA) as a function