Skip to Content
Merck
All Photos(1)

Documents

1.13203

Millipore

UV lamp

Handheld, Portable, 366 nm, Suitable for microbiology

Synonym(s):

Ultraviolet lamp, Ultraviolet ray lamp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
39101616
NACRES:
NB.14

Quality Level

description

SimPlate™ Total Plate Count (66003BC), SimPlate™ Yeast & Mold (66004BC), SimPlate™ Total Coliform & E. coli CI (66008BC)

packaging

pkg of (1 unit)

technique(s)

thin layer chromatography (TLC): suitable

application(s)

food and beverages
microbiology
sample preparation

storage temp.

15-25°C

General description

The UV lamp emits ultraviolet wavelengths around 366 nm. It can be used for the detection of Escherichia coli by means of MUG cleavate in a culture medium containing MUG (Flurocult or Chromocult® media) or in conjunction with Bactident E. coli (1.13303). Our UV lamp allows the professionals to work more quickly, efficiently, and reliably.

Application

UV lamp (366 nm) is used for the detection of fluorescence in laboratory applications such as chromatography, microbiology, or other areas of analysis.

Legal Information

BACTIDENT is a trademark of Merck KGaA, Darmstadt, Germany
CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service