Z305170
Duran® laboratory bottles, with caps
capacity 100 mL, blue PP screw cap and pouring ring
Synonym(s):
218012458, Bottles with Caps, Duran Bottles
Select a Size
$308.00
Estimated to ship onMarch 24, 2025
Select a Size
About This Item
$308.00
Estimated to ship onMarch 24, 2025
Recommended Products
material
borosilicate glass 3.3 bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GL45)
sterility
non-sterile
feature
graduations
with cap
packaging
pack of 10 ea
manufacturer/tradename
DURAN® 21 801 24 5
capacity
100 mL
diam. × H
56 mm × 105 mm
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Complies with the standards in ISO 4796-1:2016 Laboratory glassware -- Bottles -- Part 1: Screw-neck bottles (does not apply to 10 ml bottle)
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- Retrace Code, and certificate for manufacturing lot traceability
- Fired-on, highly durable white print
- Suitable for long-term storage
- Easy-to-read scale with graduation marks to estimate the content
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service