Key Documents
SYNF301410
Synthware single-neck round-bottom flask
10 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F301420, glass round flask
Select a Size
1 EA
$24.60
$24.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$24.60
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$24.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301410
capacity
10 mL
10 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)