Key Documents
Z114235
Supelco
Hand-operated aluminum cap crimper
O.D. 13 mm
Select a Size
1 EA
$552.00
$552.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$552.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
$552.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
O.D.
13 mm
General description
  • Used to attach aluminum seals to bottles and vials with a crimp/serum finish
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Polished crimping jaws provide consistent sealing
  • 2 year warranty
Choose from one of the most recent versions:
Certificates of Analysis (COA)