Key Documents
Z114227
Supelco
Hand-operated aluminum cap crimper
O.D. 11 mm
Select a Size
1 EA
$570.00
$570.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$570.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
$570.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
O.D.
11 mm
General description
  • Used to attach aluminum seals to bottles and vials with a crimp/serum finish
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Polished crimping jaws provide consistent sealing
  • 2 year warranty
Other Notes
Includes five inserts to provide a double-aggressive liner seal when used with open-top seals.
Choose from one of the most recent versions:
Certificates of Analysis (COA)