Key Documents
Z166006
Sigma-Aldrich
Wheaton open-top seals
size 13 mm, rubber septum
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
rubber septum
feature
closure type crimp seal
size
13 mm
fitting
closure for 13 mm crimp top vial
General description
It is used as an air-tight seal, and prevents contamintion
With PTFE-faced septum.
Choose from one of the most recent versions:
Certificates of Analysis (COA)